Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Vend Pack Cut Resistant Medium Duty Glove

Catalog No. 17985335 Shop All Ansell Products
Change view
Click to view available options
Size:
5
6
7
8
9
10
11
12
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-985-335 5
17-985-336 6
17-985-337 7
17-985-338 8
17-985-339 9
17-985-340 10
17-985-341 11
17-985-342 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-985-335 Supplier Ansell™ Supplier No. 48707VP050
Add to Cart
Add to Cart

Affordable safety gloves offering superb grip

  • EDGE™ 48-707 cut and abrasion-resistant gloves provide ANSI cut level A6 and EN 388 ISO cut level F, which afford a very high level of cut protection
  • Owing to their palm PU coating, these protective gloves combine good grip as well as dexterity, and are suitable for dry or slightly oily environments
  • In addition, EDGE™ 48-707 gloves meet electrostatic discharge protection needs and feature anti-static capabilities
  • These grey hand protection gloves also possess a seamless liner and knitted wrist, which deliver a secure fit for use in applications that require high cut resistance
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 388
Certifications/Compliance (Europe) Ecovadis Label
Coated Area Palm
Construction Knit
Cuff Material HPE
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A6
Hand Specific
Latex Free Yes
Material HPPE
Product Type Mechanical Protection Gloves
Size 5
EN388 Standard EN388:4X43F
For Use With (Application) Aeronautics, Assembly Line
Gender Unisex
Glove Cuff Color Gray
Glove Cuff Style Knitwrist
Grip Design Rough
Liner Material Glass Fiber
Lining Knit Lining
Packaging Type Bag
Quantity 12 Pairs
Thumb Style Keystone
Yarn Gauge 13
Length (English) 11.02 in.
Length (Metric) 280 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.