Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Venezuelan Equine Encephalitis Virus NSP1 Antibody (HL1593) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325737
Description
Venezuelan Equine Encephalitis Virus NSP1 Monoclonal antibody specifically detects Venezuelan Equine Encephalitis Virus NSP1 in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Venezuelan Equine Encephalitis Virus NSP1 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HL1593 | |
Western Blot 1:5000-1:50000, Immunohistochemistry, Immunohistochemistry-Paraffin | |
mRNA-capping enzyme nsp1, VEEV Non-Structural Protein 1, VEEV NSP1, Venezuelan Equine Encephalitis Virus Non-Structural Protein 1 | |
Recombinant protein encompassing a sequence within the center region of Venezuelan Equine Encephalitis Virus NSP1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction