Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Venezuelan Equine Encephalitis Virus NSP1 Recombinant Rabbit Monoclonal Antibody (HL1593)
GREENER_CHOICE

Catalog No. PIMA547105
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547105 Supplier Invitrogen™ Supplier No. MA547105
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

Venezuelan equine encephalitis virus (VEEV) is a member of genus Alphavirus in the family Togaviridae VEEV encodes four nonstructural proteins (nsP1, nsP2, nsP3 and nsP4) and five structural proteins (capsid (C), envelope (E) 3, E2, 6k, and E1) The Nsp1 gene of VEEV contains a 51-nt conserved sequence element (CSE) that is essential for virus replication Mutation or deletion of CSE results in loss of infectivity of VEEV, probably due to inefficient recognition of the viral RNA core promoter element by the virus replication complex Nsp1 functions as a methyltransferase and guanylylation enzyme, two properties needed for the 5' capping of VEEV genomic RNA [PMID: 30781656].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Venezuelan Equine Encephalitis Virus NSP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1593
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias VEEV NSP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of Venezuelan equine encephalitis virus nsp1. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.