Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Venezuelan Equine Encephalitis Virus NSP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144447
Description
Venezuelan Equine Encephalitis Virus NSP1 Polyclonal Antibody for Western Blot, ELISA
Venezuelan equine encephalitis virus (VEEV) is a member of genus Alphavirus in the family Togaviridae VEEV encodes four nonstructural proteins (nsP1, nsP2, nsP3 and nsP4) and five structural proteins (capsid (C), envelope (E) 3, E2, 6k, and E1) The Nsp1 gene of VEEV contains a 51-nt conserved sequence element (CSE) that is essential for virus replication Mutation or deletion of CSE results in loss of infectivity of VEEV, probably due to inefficient recognition of the viral RNA core promoter element by the virus replication complex Nsp1 functions as a methyltransferase and guanylylation enzyme, two properties needed for the 5' capping of VEEV genomic RNA [PMID: 30781656].
Specifications
Venezuelan Equine Encephalitis Virus NSP1 | |
Polyclonal | |
Unconjugated | |
VEEV NSP1 | |
Recombinant Venezuelan equine encephalitis virus (strain Trinidad donkey) (VEEV) Non-structural polyprotein (1-288aa). | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
1.366 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction