Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Venezuelan Equine Encephalitis Virus NSP4 Recombinant Rabbit Monoclonal Antibody (HL1741)
GREENER_CHOICE

Catalog No. FEMA550330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550330 Supplier Invitrogen™ Supplier No. MA550330
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Venezuelan Equine Encephalitis Virus NSP4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

Venezuelan Equine Encephalitis Virus (VEEV) is a mosquito-borne virus that can cause severe illness in both humans and animals. The virus is classified as a category B bioterrorism agent by the Centers for Disease Control and Prevention (CDC) due to its potential use as a biological weapon. The NSP4 gene of the VEEV is a non-structural protein that plays a crucial role in viral replication and pathogenesis. NSP4 is a promising target for the development of antiviral drugs and vaccines against VEEV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Venezuelan Equine Encephalitis Virus NSP4
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1741
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias VEEV NSP4
Host Species Rabbit
Immunogen Recombinant fragment of Venezuelan equine encephalitis virus nsP4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.