Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Venture Gear VG80 Series Ear Muffs

Catalog No. 19555711 Shop All Pyramex Safety Products Products
Click to view available options
Color:
Black graphite pattern
Gray
High-visibility Lime
High-visibility Orange
Pink
Purple
Teal

Venture Gear VG80 series of ear muffs are comfortable and stylish with a variety of colors to choose from.

  • NRR 26 dB increased to 36 dB when worn with ear plugs
  • Low-profile design
  • Height adjustment to fit all sizes
  • Fold-away padded headband

Specifications

Color High-visibility Orange
ANSI S3.1974 Yes
Cushion Color Black
For Use With (Application) Hearing Protection
High Visibility Yes
Model VG80 Series
Packaging Type Clamshell
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 26 dB
Headband Color Black
Dielectric No
Style Over-the-head
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.