Learn More
Pyramex™ VentureGear Tactical™ Drone™ Safety Eyewear
Catalog No. p-7084351 Shop All Pyramex Safety Products Products
Click to view available options
Lens Tint:
Clear
Gray
This item is not returnable. View return policy
This item is not returnable. View return policy
Packaged in a VentureGear Tactical™ branded box with temple tag for counter top or pegboard display
  • Half-frame design
  • Stylish, dual injected temples
  • Adjustable rubber nosepiece
  • Anti-fog, scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Certifications/Compliance ANSI Z87.1,CAN/CSA Z94.3-07
    Lens Coating Scratch-resistant
    Lens Tint Clear
    Frame Color Black
    Lens Material Polycarbonate
    Frame Type Half frame with ventilation
    Product Content Correction
    Your input is important to us. Please complete this form to provide feedback related to the content on this product.
    Product Title
    Your feedback has been submitted: Thank you for helping us improve our website.