Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ VentureGear™ Waverton Safety Eyewear

Packaged in a VentureGear™ branded polybag with retail tag

$269.60 - $441.00

Specifications

Certifications/Compliance ANSI Z87.1
Weight (Metric) 24.4 g
Lens Material Polycarbonate
Frame Material Polycarbonate
View More Specs

Products 5
Catalog Number Mfr. No. Frame Color Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Frame Color Lens Tint Price Quantity & Availability  
19-156-027
View Documents
Pyramex™
VGSB1126DB
Black Green
Case of 12 for $269.60
Only null left
Add to Cart
 
19-156-028
View Documents
Pyramex™
VGSB1165DB
Black Blue, Mirror
Case of 12 for $299.78
Only null left
Add to Cart
 
19-156-029
View Documents
Pyramex™
VGSB1170DB
Black Silver, Mirror
Case of 12 for $441.00
Only null left
Add to Cart
 
19-156-060
View Documents
Pyramex™
VGSBR1118DB
Brown Bronze
Case of 12 for $269.60
Only null left
Add to Cart
 
19-156-061
View Documents
Pyramex™
VGSBR1156DB
Brown Mirror
Case of 12 for $299.78
Only null left
Add to Cart
 
Description

Description

  • Flexible rubber nosepiece
  • Dual-injected straight-back temples
  • Scratch resistant polycarbonate lens provides 99% UVA/B/C protection
  • Specifications

    Specifications

    ANSI Z87.1
    24.4 g
    Polycarbonate
    Polycarbonate
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.