Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VENTX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578478
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578478 Supplier Invitrogen™ Supplier No. PA578478
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Raji, NIH3T3 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

VENTX is a member of the Lymphoid-enhancing factor/T-cell factors (LEF1/TCF), which are a high-mobility group of transcriptional factors that play essential roles in cell fate determination during early embryogenesis and ontogenesis. VENTX is a member of the Vent family of homeodomain proteins. It is a novel LEF/TCF-associated transcriptional modulator. VENTX is predominately expressed in hematopoietic cells. VENTX may function as a transcriptional repressor and be involved in mesodermal patterning and hemapoietic stem cell maintenance, and has been suggested to have a potential role in the hematopoietic malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VENTX
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Ventx
Gene Accession No. O95231
Gene Alias hemopoietic progenitor homeobox protein VENTX2; homeobox protein VENTX; HPX42B; NA88A; VENT homeobox; VENT homeobox homolog; VENT-like homeobox 2; VENT-like homeobox protein 2; VENTX; VENTX2
Gene Symbols Ventx
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human VENTX. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110333234, 27287
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.