Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VEPH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121206
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121206 Supplier Invitrogen™ Supplier No. PA5121206
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: U-87MG, 293T, HT-1080, Mouse brain. The target is usually found in the following locations: Cell membrane, Cytoplasmic side, Peripheral membrane protein. Immunogen sequence: KSLAFHTMLT KGVGSDDGED ENRGDIPASI SLSEIDPLGQ GNDKLPFKTD TERSQLGESS VSYPNIIHID SENLSETVKE NSQEETPETT ASPIEYQDKL.

Ventricular zone-expressed PH domain-containing protein homolog 1 (VEPH1), also known as KIAA1692 or Protein melted, is a 833 amino acid protein belonging to the MELT/VEPH family. VEPH1 is a peripheral membrane protein localized to the cytoplasmic side. VEPH1 has one PH domain which is required for the membrane targeting of the protein. The human gene encoding VEPH1 is 76% homologous to the Drosophila VEPH1 gene which encodes a protein that acts as a regulator of the insulin signaling pathway. It is thought that this role of VEPH1 is conserved across species. VEPH1 exists as three isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEPH1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene VEPH1
Gene Accession No. A1A535, Q14D04
Gene Alias 2810471M23Rik; KIAA1692; MELT; Protein melted; protein melted homolog; RGD1308217; ventricular zone expressed PH domain containing 1; ventricular zone expressed PH domain homolog 1; ventricular zone expressed PH domain-containing 1; ventricular zone-expressed PH domain-containing protein 1; Ventricular zone-expressed PH domain-containing protein homolog 1; VEPH; VEPH1
Gene Symbols VEPH1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 448-547 of human VEPH1 (NP_078897.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72789, 79674
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.