Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VEPH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536006
Description
Recommended positive controls: U87-MG, SK-N-SH, IMR-32, SK-N-AS, mouse heart. Predicted reactivity: Mouse (91%), Rat (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ventricular zone-expressed PH domain-containing protein homolog 1 (VEPH1), also known as KIAA1692 or Protein melted, is a 833 amino acid protein belonging to the MELT/VEPH family. VEPH1 is a peripheral membrane protein localized to the cytoplasmic side. VEPH1 has one PH domain which is required for the membrane targeting of the protein. The human gene encoding VEPH1 is 76% homologous to the Drosophila VEPH1 gene which encodes a protein that acts as a regulator of the insulin signaling pathway. It is thought that this role of VEPH1 is conserved across species. VEPH1 exists as three isoforms produced by alternative splicing.
Specifications
VEPH1 | |
Polyclonal | |
Unconjugated | |
VEPH1 | |
2810471M23Rik; KIAA1692; MELT; Protein melted; protein melted homolog; RGD1308217; ventricular zone expressed PH domain containing 1; ventricular zone expressed PH domain homolog 1; ventricular zone expressed PH domain-containing 1; ventricular zone-expressed PH domain-containing protein 1; Ventricular zone-expressed PH domain-containing protein homolog 1; VEPH; VEPH1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
72789, 79674 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
A1A535, Q14D04 | |
VEPH1 | |
Recombinant fragment corresponding to a region within amino acids 15 and 274 of VEPH1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction