Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Verapamil hydrochloride
GREENER_CHOICESDP

Catalog No. 06541G Shop All Novus Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-541-G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 06-541-G Supplier R&D Systems™ Supplier No. 0654/1G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. CaV1.x blocker

Verapamil hydrochloride is an L-type calcium channel blocker. Vasodilator and adrenergic antagonist. Verapamil induces autophagy by modulating cellular metabolism. Verapamil has been identified as targeting human host proteins that interact with SARS-CoV-2.

Chemical Identifiers

CAS 152-11-4
Molecular Formula C27H39ClN2O4

Specifications

Quantity 1 g
Product Line Tocris
Formula Weight 491.07
Purity 0.98
Molecular Weight (g/mol) 491.069
Chemical Name or Material Verapamil hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.