Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Verapamil Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607705
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle

V1072, 152-11-4, C27H38N2O4·HCl

Spectrum™ Chemical Verapamil Hydrochloride, USP belongs to a class of drugs known as calcium channel blockers and is used to treat high blood pressure. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 152-11-4
Molecular Formula C27H39ClN2O4
Molecular Weight (g/mol) 491.07
InChI Key DOQPXTMNIUCOSY-UHFFFAOYNA-N
IUPAC Name hydrogen 2-(3,4-dimethoxyphenyl)-5-{[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino}-2-(propan-2-yl)pentanenitrile chloride
SMILES [H+].[Cl-].COC1=CC=C(CCN(C)CCCC(C#N)(C(C)C)C2=CC=C(OC)C(OC)=C2)C=C1OC

Specifications

CAS Max % 1
pH 4.5 to 6.5
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.