Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Verbascoside 97.0+%, TCI America™
SDP

Catalog No. p-7137946
Click to view available options
Quantity:
10 mg
50 mg

Chemical Identifiers

CAS 61276-17-3
Molecular Formula C29H36O15
Molecular Weight (g/mol) 624.59
MDL Number MFCD00221751,MFCD00221751
InChI Key FBSKJMQYURKNSU-ZLSOWSIRSA-N
Synonym Acteoside, Kusaginin, 2-(3,4-Dihydroxyphenyl)ethyl 3-O-(6-Deoxy-alpha-L-mannopyranosyl)-4-O-[(2E)-3-(3,4-dihydroxyphenyl)-2-propenoyl]-beta-D-glucopyranoside
PubChem CID 45358104
IUPAC Name (2R,3R,4R,5R,6R)-6-[2-(3,4-dihydroxyphenyl)ethoxy]-5-hydroxy-2-(hydroxymethyl)-4-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-3-yl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](OCCC3=CC=C(O)C(O)=C3)O[C@H](CO)[C@H]2OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@H](O)[C@H](O)[C@H]1O

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 624.59
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Verbascoside
RTECSNumber LZ5786000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.