Description
- Available with or without a critical orifice
- Interchangeable critical orifice
Order Info
Order orifices for alternate sampling times, or replacement orifices, separately.
Specifications
Specifications
| Material | Stainless Steel |
| Size (English) Orifice | 0.0016 in. |
| Size (Metric) Orifice | 0.004 cm |
| Type | Veriflo SC423 XL Flow Controller |