Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ VeriShield™ Dielectric Multi-Position Ear Muffs

Catalog No. 19085483 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
CSA Z94.2:
Class A
Class AL
Noise Reduction Rating (NRR):
25 dB
30 dB
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. CSA Z94.2 Noise Reduction Rating (NRR)
19-085-483 Class AL 30 dB
19-085-539 Class A 25 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-085-483 Supplier Honeywell™ Supplier No. 1035187VS
Add to Cart
Add to Cart

VeriShield™ Dielectric Multi-Position Ear Muffs can be adjusted and worn in three different positions depending on comfort and application.

  • Multi-Position comfortably fits a wide range of head sizes
  • Wide cup opening is ideal for workers with large ears, or who wear hearing aids
  • Lightweight cushioned headband
  • Memory foam ear cushions
  • Oil-resistant cup surface withstands heavy-work environments and frequent cleaning
  • Features Sound Management Technology (SMT)
  • Available in mid or high level attenuation

    Specifications

    Color Black
    ANSI S3.1974 Yes
    CSA Z94.2 Class AL
    Cushion Color Black
    For Use With (Application) Construction, Electrical, Manufacturing, Oil and Gas, Mining, Steel and Metals, Agriculture
    High Visibility No
    Model VS130M
    Pairs per Package 1 Ea.
    Noise Reduction Rating (NRR) 30 dB
    Product Type VeriShield
    Dielectric Yes
    Material ABS
    Style Multi-position
    Foldable No
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.