Learn More
Honeywell™ VeriShield™ 100 Series Over-the-Head Ear Muffs
VeriShield™ Over-the-Head style ear muffs feature air flow control technology for maximum attenuation without increasing pressure on the head.
Supplier: Honeywell™ 1035106VS
This item is not returnable.
View return policy
Description
- Comfortably fits a wide range of head sizes
- Wide cup opening is ideal for workers with large ears, or who wear hearing aids
- Lightweight cushioned headband
- Memory foam ear cushions
- Oil-resistant cup surface withstands heavy-work environments and frequent cleaning
- Features Sound Management Technology (SMT) which improves employee safety by blocking harmful noise while allowing voices and signals to be heard more naturally
- Designed for use with other PPE
Specifications
High-visibility Yellow | |
Class AL | |
VS120FHV | |
27 dB | |
No | |
Over-the-head |
Yes | |
Construction, Manufacturing, Oil and Gas, Mining, Steel and Metals, Welding, Agriculture | |
1 Ea. | |
VeriShield | |
ABS | |
Yes |