Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ VeriShield™ 100 Series Hard Hat Mount Ear Muffs

Catalog No. 19085635 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Black
High-visibility Yellow
CSA Z94.2:
Class A
Class AL
Class B
Dielectric:
No
Yes
Noise Reduction Rating (NRR):
21 dB
23 dB
24 dB
25 dB
27 dB
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color CSA Z94.2 Dielectric Noise Reduction Rating (NRR)
19-085-635 High-visibility Yellow Class AL No 27 dB
19-085-636 Black Class B No 21 dB
19-085-537 Black Class A No 24 dB
19-085-538 Black Class AL No 27 dB
19-085-484 Black Class A Yes 23 dB
19-085-485 Black Class A Yes 25 dB
19-085-486 Black Class AL Yes 27 dB
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-085-635 Supplier Honeywell™ Supplier No. 1035127VS
Add to Cart
Add to Cart

VeriShield™ Hard Hat Ear Muffs mount to a variety of helmets and ensures protection against noise levels while allowing voices and signals to be heard more naturally.

  • Helmet mounted versions
  • Wide cup opening ideal for workers with large ears, or who wear hearing aids
  • Lightweight cushioned headband
  • Memory foam ear cushions
  • Oil-resistant cup surface withstands oily, heavy-work environments and frequent cleaning
  • Features Sound Management Technology (SMT) allows voices and signals to be heard more naturally
  • Designed for use with other PPE
  • Dielectric (models 19-085-484, 19-085-485, 19-085-486 only)

Specifications

Color High-visibility Yellow
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Cushion Color Black
For Use With (Application) Construction, Manufacturing, Oil and Gas, Mining, Steel and Metals, Agriculture
High Visibility Yes
Model VS130HHV
Pairs per Package 1 Ea.
Noise Reduction Rating (NRR) 27 dB
Dielectric No
Style Hard Hat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.