Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ Ammonium Phosphate Matrix Modifier for AAS 20,000 ppm in 1% HNO3, Ricca Chemical

Click to view available options
Quantity:
50 mL
Description
- 20,000 ppm in 1% HNO3
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
Chemical Name or Material | Ammonium Phosphate AAS Matrix Modifier |
Packaging | Natural Poly Bottle |
CAS | 7732-18-5, 7722-76-1, 7697-37-2 |
CAS Min % | 95.06%, 1.96%, 0.98% |
CAS Max % | 98.94%, 2.04%, 1.02% |
Specific Gravity | 1.01 |
Physical Form | Liquid |
Color | Colorless |
Melting Point | Approximately 0°C |
For Use With (Equipment) | Atomic Absorption Spectrophotometer |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction