Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Barium Standard for ICP 10 g/L in 2% HNO3, Ricca Chemical
SDP

Catalog No. p-7073846 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010438100 100 mL
RV010438500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010438100 Supplier Ricca Chemical Company Supplier No. RV010438100N
Only null left
Add to Cart
Add to Cart

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 10 g/L in 2% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10022-31-8 , 7697-37-2 , 7732-18-5
Molecular Formula BaN2O6
Molecular Weight (g/mol) 261.34
MDL Number MFCD00003442
InChI Key IWOUKMZUPDVPGQ-UHFFFAOYSA-N
PubChem CID 24798
IUPAC Name barium(2+) dinitrate
SMILES [Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 94.18%, 1.96%, 1.86%
CAS Max % 98.02%, 2.04%, 1.94%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.