Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ Formate - HCOO- Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
100 mL
250 mL
500 mL
Description
- 1000 ppm in H2O
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
Packaging | Natural LDPE Bottle |
CAS Min % | 97.85%, 0.15% |
CAS Max % | 100%, 0.15% |
Specific Gravity | 1.01 |
Physical Form | Liquid |
Melting Point | Approximately 0°C |
Quantity | 100 mL |
Grade | High Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction