Learn More
VeriSpec™ Gold Standard for ICP 1000 ppm in 2% HCl, Ricca Chemical
Shop All Ricca Chemical Company Products

Description
- 1000 ppm in 2% HCl
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS Min % | 95.87%, 1.96%, 0.17% |
| CAS Max % | 99.79%, 2.04%, 0.17% |
| Specific Gravity | 1 |
| Physical Form | Liquid |
| Melting Point | Approximately 0°C |
| Type | Standard |
| Quantity | 250 mL |
| Grade | High Purity |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.