Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ ICP Calibration Standard 15, Ricca Chemical
SDP

Catalog No. p-7073950
Click to view available options
Quantity:
100 mL
500 mL

5 ppm: Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se Sr, Zn; 50 ppm K /nManufactured and Tested in

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS Min % 4.90%, 0.01%, 0.00%, 0.00%, 0.00%, 0.00%
CAS Max % 5.10%, 0.01%, 0.00%, 0.00%, 0.00%, 0.00%
CAS 13473-90-0, 13548-38-4, 10141-05-6, 10325-94-7, 10022-31-8, 7697-37-2
Specific Gravity 1.07
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.