Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ Instrument Calibration Standard 4, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
100 mL
500 mL
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| CAS Min % | 4.90%, 2.99%, 2.01%, 1.81%, 1.26% |
| CAS Max % | 5.10%, 3.11%, 2.09%, 1.89%, 1.32% |
| CAS | 10377-60-3, 10124-37-5, 7631-99-4, 7757-79-1, 7697-37-2 |
| Specific Gravity | 1.06 |
| Physical Form | Liquid |
| UN Number | UN3264 |
| Melting Point | Approximately 0°C |
| Quantity | 100 mL |
| Grade | High Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction