Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ Nexion ICP-MS Calibration Standard for Set Up 8, Ricca Chemical

Click to view available options
Quantity:
500 mL
Description
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
Packaging | Natural Poly Bottle |
CAS Min % | 4.90%, 1.96%, 0.91% |
CAS Max % | 5.10%, 2.04%, 0.95% |
CAS | 7790-69-4, 7697-37-2, 7647-01-0, 7732-18-5 |
Specific Gravity | 1.06 |
Physical Form | Liquid |
UN Number | UN3264 |
Melting Point | Approximately 0°C |
Quantity | 500 mL |
Grade | High Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction