Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Nitrite - NO2- Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
SDP

Catalog No. p-7074056 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
100 mL
250 mL
500 mL

Manufactured and Tested in an ISO 17025/Guide 34 Facility

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7632-00-0, 7732-18-5
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Packaging Natural LDPE Bottle
CAS Min % 97.85%, 0.15%
CAS Max % 100%, 0.15%
Specific Gravity 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.