Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Palladium Nitrate Matrix Modifier for AAS 2,000 ppm in 1% HNO3, Ricca Chemical
SDP

Catalog No. RV01013450N Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
50 mL

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Chemical Name or Material Palladium Nitrate AAS Matrix Modifier
Packaging Natural Poly Bottle
CAS 7732-18-5, 7697-37-2, 10102-05-3
CAS Min % 96.82%, 0.98%, 0.20%
CAS Max % 100%, 1.02%, 0.20%
Specific Gravity 1.01
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) Atomic Absorption Spectrophotometer
Solubility Miscible with water
Solution Type Calibration Standard
Type Matrix Modifier
Quantity 50 mL
Grade High Purity
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.