Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VeriSpec™ QC Standard 23, Ricca Chemical

Click to view available options
Quantity:
100 mL
Description
- 1000 ppm: Ag, Al, B, Ba, Bi, Ca, Cd, Co, Cr, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Sr, Tl, Zn
- Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
- Product accompanied with detailed Certificate of Analysis
Specifications
Specifications
Packaging | Natural LDPE Bottle |
CAS Min % | 9.8%, 1.3622%, 0.1568% |
CAS Max % | 10.2%, 1.4178%, 0.1632% |
CAS | 13473-90-0, 7761-88-8, 7697-37-2 |
Specific Gravity | 1.09 |
Physical Form | Liquid |
UN Number | UN2031 |
Quantity | 100 mL |
Grade | High Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction