Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Silicon Standard for ICP 10 g/L in H2O, Ricca Chemical
SDP

Catalog No. RV010415100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010415100 100 mL
RV010415500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010415100 Supplier Ricca Chemical Company Supplier No. RV010415100N

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 10 g/L in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 16919-19-0 , 7732-18-5
Molecular Formula F6H8N2Si
Molecular Weight (g/mol) 178.153
InChI Key ITHIMUMYFVCXSL-UHFFFAOYSA-P
PubChem CID 28145
IUPAC Name diazanium;hexafluorosilicon(2-)
SMILES [NH4+].[NH4+].F[Si-2](F)(F)(F)(F)F

Specifications

Packaging Natural LDPE Bottle
CAS Min % 91.79%, 6.21%
CAS Max % 95.53%, 6.47%
Specific Gravity 1
Physical Form Liquid
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.