Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ VeritiPro™ Thermal Cycler, 384-well

Catalog No. A47399
Encompass_Preferred
Change view
Click to view available options
Product Type:
Thermal Cycler
Thermal Cycler with Plastic Package
Warranty:
3 years of total service coverage
3 years of total service coverage with instrument rapid exchange
Limited Warranty
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Warranty
A47399 Thermal Cycler with Plastic Package Limited Warranty
A48140 Thermal Cycler Limited Warranty
A50327 Thermal Cycler with Plastic Package 3 years of total service coverage
A50328 Thermal Cycler with Plastic Package 3 years of total service coverage with instrument rapid exchange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A47399 Supplier Applied Biosystems™ Supplier No. A47399
Add to Cart
Add to Cart
Request a Quote

The Applied Biosystems VeritiPro Thermal Cycler delivers proven reliability with advanced temperature control technology and connectivity.

The Applied Biosystems VeritiPro Thermal Cycler delivers proven reliability with advanced temperature control technology and connectivity. Take advantage of next-level PCR technology featuring faster ramping speeds and lower noise output. Connect to the cloud-enabled VeritiPro Thermal Cycler remotely. Conveniently design and securely upload your methods, monitor runs, and check instrument availability from any mobile device or desktop computer using Connect, our cloud-based platform.

This system may be purchased with a plastics package, extended warranty, or both depending on lab needs.

Features of the VeritiPro Thermal Cycler include:

Remote capabilities

By connecting your VeritiPro Thermal Cycler to Connect, you will have complete control of your instrument while away from the lab. With Connect you can check instrument availability through the mobile app, download experimental report files, receive alerts, and more. Download our Instrument Connect App, updated with this functionality and visit Connect to design and securely upload your methods from any mobile device or desktop computer.

Simulation mode

The VeritiPro Thermal Cycler is equipped with simulation modes, allowing it to be run using the same ramp rates as previous or different instruments. With this feature you will be able to mimic the ramp rates of the Applied Biosystems 2720, Bio-Rad T100, Bio-Rad MyCycler, Bioer XP Cycler, Eppendorf Mastercycler, MJ Research PTC-200, and Takara Dice PCR thermal cyclers.

Our quietest instrument yet

With all the equipment running in the lab, why should your thermal cycler add to the noise level? We have revised the internal instrument technology to provide your lab with a thermal cycler that runs at a noise level of less than 48 dB(A).

8-inch touch-screen control

The improved, intuitive, and responsive touch-screen interface allows you to program with ease and limits the amount of time spent training new users.

Compact footprint

With the increasing popularity of our smaller instruments, we have updated our trusted Veriti Thermal Cycler with a smaller footprint. The VeritiPro Thermal Cycler can fit on just about any workbench with its dimensions of 24.5 cm wide and 46.7 cm deep.

Plastics

Applied Biosystems PCR plastics

Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for any experiment.

MicroAmp EnduraPlates

MicroAmp EnduraPlates offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. They are a solution for work that requires special handling and an even greater degree of durability for use with multi-instrument experiments. They are validated for use with Applied Biosystems PCR thermal cyclers for optimal fit and performance.

  • Optimized for performance with Applied Biosystems instruments
  • Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
  • Constructed to ANSI/SBS standard, well-suited to high-throughput laboratories and robot handling
  • Available in multiple colors for easy organization of laboratory assays
  • Excellent mechanical stability and flatness for your complete workflow needs

MicroAmp Clear Adhesive Films

MicroAmp Clear Adhesive Films create a secure seal across an entire microplate to prevent evaporation and well-to-well contamination.

  • Adhesive provides tight seal over wells to prevent leaks and evaporation
  • Easy to apply and remove
  • Available as Clear (economical and sticky) or Optical (ready for real-time PCR and non-tacky)

Warranties

Rapid Exchange (REX) extended warranty

The package offered here provides additional warranty coverage to the standard two-year warranty. With the Rapid Exchange (REX) extended warranty, if your instrument needs repair, just call our Support Center and we will exchange it for a factory-certified refurbished replacement instrument within one business day from the time we determine a replacement is required. Instruments are typically received within three days of shipment and are yours to keep. All parts, labor, and shipping charges are included, and existing warranty contracts will be transferred to replacement units.

The Rapid Exchange extended warranty is ideal for customers with user-installable instruments that do not require asset tag consistency, and for those operating under critical timelines.

AB Repair Center (ABRC) extended warranty

The package offered here provides additional warranty coverage to the standard two-year warranty. With ABRC support, factory-trained engineers will perform maintenance or repair your instrument at our remote service center. Just send us your instrument and we will maintain or repair it and ship it back to you typically within three weeks. All parts, labor, and shipping charges are included.

The ABRC extended warranty is ideal for customers in regulated environments who must maintain asset tag consistency to comply with regulated protocols.

Certain exclusions apply. Contact your Service and Support Representative for details.

TRUSTED_SUSTAINABILITY

Specifications

Thermal Accuracy ±0.25°C (35°C to 99.9°C)
Thermal Uniformity <0.5°C (20 sec. after reaching 95°C)
Dimensions 18.3 x 9.6 x 8.5 in. (46.5 x 24.5 x 21.7 cm)
Capacity 384-well
Voltage 100/240 V
Display Type 8 in. Color TFT LCD Touchscreen
Product Type Thermal Cycler with Plastic Package
Warranty Limited Warranty
Thermal Range 0°C to 100°C
User Interface USB, Wi-Fi, Ethernet
Wattage 700 W
Max. Ramp Rate 5°C/sec.
No. of Programs >1000
Description VeritiPro™ Thermal Cycler + Plastics Package, 384-well, EnduraPlates
Quantity 1 Package
Weight 26.5 lb. (12 kg)
Block Configurations 384-well Isothermal Block
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.