Learn More
Vernier Basic ProScope HR2 Kit
SDPSCI_ED
Catalog No. s16092nd Shop All Vernier Software Products
Click to view available options
Description:
ProScope HR 2 Kit with 50X Lens
High-resolution handheld digital microscope for capturing images, videos and time-lapse photographs.
Ideal for capturing high resolution microscopic images of hair follicles, microwriting on dollar bills, and more with 50X lens.
Basic kit includes:
  • ProScope HR2 base
  • ProScope HR2 software
  • 50X lens
Proscope HR2 Specifications:
  • Image sensor: 1/3 i inch high resolution 2.0 megapixel sensor
  • Connection type: USB
Image capture resolutions:
  • 320x240 pixels
  • 640x480 pixels
  • 1280x1024 pixels
  • 1600x1200 pixels
Video capture resolutions:
  • 320x240 pixels @ 30 fps
  • 640x480 pixels at @ 15 fps
  • 1280x1024 pixels at @ 3.75 fps
  • 1600x1200 pixels at @ 2.5 fps
Specifications
Description ProScope HR 2 Kit with 50X Lens
Includes ProScope HR2 base, ProScope HR software, 50X lens
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.