Learn More
Vernier Replacement Cuff for Blood Pressure Sensor
SDPSCI_ED
Catalog No. p-5164621 Shop All Vernier Software Products
Change view
Click to view available options
Size:
Large
Small
Standard
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S16135ND Standard
S16005ND Large
S16004ND Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S16135ND Supplier Vernier Supplier No. CUFFSTD
Only null left
Add to Cart
Add to Cart
Includes
One replacement cuff for use with Vernier Blood Pressure Sensor (S16101ND).
Blood Pressure Sensor Cuff for use with Vernier Blood Pressure Sensor (S16101ND).
  • For students with different arm circumferences
  • Small: 18 to 27cm
  • Standard: 27 to 39cm
  • Large: Greater than 39cm and less than 50cm
Specifications
Size Standard
For Use With (Equipment) Vernier Blood Pressure Sensor (S16101ND)
Product Type Blood Pressure Cuff
WARNING: Cancer - www.P65Warnings.ca.gov