Learn More
Vernier Vernier Disposable Bacterial Filter
SDPSCI_ED
Catalog No. S16252ND Shop All Vernier Software Products
Click to view available options
:
Disposable Bacterial Filter
This item is intended for educational purposes only.
This item is intended for educational purposes only.
Significantly reduces how often you will need to sterilize your spirometer's flow head.
  • Microbacterial filter, for use with Vernier Spirometer
  • Filter's 30mm ID opening allows for an easy, air-tight fit to the flow head
  • Sold in pack of 10
Specifications
Product Type Disposable Bacterial Filter
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.