Learn More
Vernier Vernier Exercise Heart Rate Monitor
Shop All Vernier Software Products
Click to view available options
:
Exercise Heart Rate Monitor
Description
Includes
Exercise Heart Rate Strap, Polar Transmitter Module (battery included), Heart Rate Receiver, User manual
- Using the chest strap, data are wirelessly transmitted to Vernier interface using Heart Rate Receiver
- Data can be transmitted using Bluetooth™ to LabQuest™ 2 (may require additional hardware), iOS™, or Android™ devices without receiver
- Chest strap is ideal for running, cycling, and other physical activities
- Radios: Bluetooth and 5kHz RF transmission
Specifications
Specifications
| Product Type | Exercise Heart Rate Monitor |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov