Learn More
Vernier Hand Dynamometer
SDPSCI_ED
Catalog No. s16174nd Shop All Vernier Software Products
Click to view available options
:
Hand Dynamometer
Catalog No. S16174ND
Supplier: Vernier HDBTA
Only null left
Add to Cart
Add to Cart
Recommended for all grades.
  • Strain-gauge based isometric Hand Dynamometer can be used to measure grip strength, pinch strength, and to perform muscle fatigue studies
  • Can be used with other sensors (e.g., EKG Sensor) to study muscular health and activity
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.