Learn More
Vernier KidWind Blade Pitch Protractor
SDPSCI_ED
Catalog No. s04804nd Shop All Vernier Software Products
Click to view available options
:
KidWind Blade Pitch Protractor
Used for measuring blade pitch angle on KidWind turbine.
  • Recommended for college, high school, and middle school science
  • Makes blade pitch experiments much easier
  • Designed with cut-out notch to fit around blade dowel, and it has ridges on back that align with KidWind hub, which makes it perfect for reading, measuring, and comparing blade pitch angles
  • Range of measurement: 0° to 70°
Warranty and Services
5 years limited
Specifications
Product Type KidWind Blade Pitch Protractor
For Use With (Application) Measuring blade pitch angle on KidWind turbine
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.