Learn More
Vernier KidWind Drivetrain Set
Shop All Vernier Software Products
Click to view available options
:
Drivetrain set
Description
Includes
8 in. hex shaft for use with Advanced Wind Experiment Kit Nacelle
- Recommended for College and High School
Does not work with the Basic Wind Experiment Kit Weightlifter Nacelle
Warranty and Services
5-year limited warranty
Specifications
Specifications
Product Type | Drivetrain Set |
For Use With (Application) | To connect wind turbine hub to a nacelle |
Includes | 8 in. hex shaft for use with Advanced Wind Experiment Kit Nacelle |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov