Learn More
Vernier KidWind Power Output Board
Shop All Vernier Software Products
Click to view available options
Product Type:
Power output board
Description
- Included in Basic Wind Experiment Kit and Advanced Wind Experiment Kit and Classroom Packs
- Includes visual voltmeter, LEDs, resistors, capacitor and sound generator
- Recommended for College and High School
Specifications:
- Visual Voltmeter Input: 0-10V
- Sound Input: 0-12V
- Capacitor: 0-2.5V
- 10, 30, 51 and 100 Ohm resistor
Warranty and Services
5-year limited warranty
Specifications
Specifications
| Product Type | Power output board |
| For Use With (Equipment) | To demonstrate power output from small electrical devices |
| Includes | Visual voltmeter, LEDs, resistors, capacitor, and sound generator |