Learn More
Vernier Vernier Neon Spectrum Tube
SDPSCI_ED
Catalog No. s16020nd Shop All Vernier Software Products
Click to view available options
:
Neon Spectrum Tube
Gas tubes for qualitative study by students noting more spectral lines and unique characteristic lines. For use with the Spectrum Tube Single Power Supply and the Spectrum Tube Carousel Power Supply.
  • Permanently enclosed in plastic carriers to prevent breakage
  • No through-the-glass electrodes
  • Ultrasafe
  • Long lasting and superior to older designs
  • For Grades 9 and up
Specifications
Product Type Neon Spectrum Tube
For Use With (Equipment) Spectrum Tube Single Power Supply and the Spectrum Tube Carousel Power Supply
Material Plastic
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.