Learn More
Vernier ProScope HR Digital Microscope Kits
SDPSCI_ED
High-resolution handheld digital microscope for capturing images, videos, and time-lapse photographs.
Specifications
Required Accessories Windows: Windows™ XP service pack 2 or newer, Quicktime 7.0 or newer, DirectX 9 or newer, available USB 2 port, Pentium 4 or equivalent; Macintosh: Mac OS 10.4.8 or newer, Quicktime 7.0 or newer, factory-installed USB 2 port, G4, G5, or Intel processor; Chromebook: Camera app and available USB 2 port
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
S16091ND
SDPSCI_ED
View Documents
Vernier
BDDLX
ProScope HR Deluxe Kit N/A N/A N/A
S16093ND
SDPSCI_ED
View Documents
Vernier
BDHRBIOL
Biology ProScope HR Kit N/A N/A N/A
Description
Description
The ProScope HR is a USB handheld digital microscope designed for use with computer. With a high-quality CMOS sensor and universal lens mount, the ProScope HR is a powerful addition to any classroom.
Biology Kit:
Use ProScope HR as dissecting microscope or attach ProScope HR to regular microscope for even more magnification.
Includes:
  • ProScope HR base
  • ProScope HR software
  • 50X lens
  • 1-10X lens
  • C-mount adapter with IR cut filter
  • Microscope lens tube adapter
  • Camera stand
  • Rugged hard-side case
Deluxe Kit:
For maximum magnification with or without regular microscope, deluxe kit has covered with its lens ensemble and accessories.
Includes:
  • ProScope HR base
  • ProScope HR software
  • 50X lens
  • 1-10X lens
  • 100X lens
  • 200X lens
  • C-mount adapter with IR cut filter
  • Microscope lens tube adapter
  • Camera stand
  • Rugged hard-side case
Proscope HR Specifications:
  • Image sensor: 1/4 in. high resolution 1.3 megapixel sensor
  • Connection type: USB
  • Image capture resolutions:
  • 320x240 pixels
  • 640x480 pixels
  • 1280x1024 pixels
  • Video capture resolutions:
  • 320x240 pixels at @30 fps
  • 640x480 pixels at @15 fps
  • 1280x1024 pixels at @3.75 fps
Specifications
Specifications
Windows: Windows™ XP service pack 2 or newer, Quicktime 7.0 or newer, DirectX 9 or newer, available USB 2 port, Pentium 4 or equivalent; Macintosh: Mac OS 10.4.8 or newer, Quicktime 7.0 or newer, factory-installed USB 2 port, G4, G5, or Intel processor; Chromebook: Camera app and available USB 2 port
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.