Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Radiation Monitor
SDPSCI_ED

Catalog No. s15994nd Shop All Vernier Software Products
Click to view available options
Sensitivity:
Alpha, Beta, Gamma, X-ray Radiation

Catalog No. S15994ND

Supplier: Vernier VRMBTD

Only null left
Add to Cart
Add to Cart

Requires

Requires a compatible interface and software to collect data. Interface and software sold separately.

Detects alpha, beta, gamma, and X-ray radiation.

  • Consists of a Geiger-Mueller tube mounted in a small, rugged, plastic case
  • Requires no battery, getting power from the data-collection interface
  • A thin window protected by a metal screen allows alpha radiation to be detected, along with beta and gamma
  • Can be used to explore radiation statistics, measure the rate of nuclear decay, and monitor radon progeny
  • Compatible interfaces: LabQuest 2, Original LabQuest, LabQuest Mini, SensorDAQ, LabPro, CBL 2, TI-Nspire Lab Cradle
  • Recommended for College and High School levels

Specifications

  • Sensor: LND 712 (or equivalent) halogen-quenched GM tube with a mica end window, 1.5 to 2.0 mg/cm2 thick
  • Sensitive to alpha, beta, gamma, and X-ray radiation
  • Powered by interface connection
  • Red LED blinks on radiation detection

Applications
Nuclear counting statistics, shielding, and decay rate measurements.

Order Info

Interface and software sold separately.

Specifications

Sensitivity Alpha, Beta, Gamma, X-ray Radiation
Product Type Radiation Monitor
Power Supply From data-collection interface
Detector Type LND 712 (or equivalent) halogen-quenched GM tube with a mica end window, 1.5 to 2.0 mg/cm2 thick
For Use With (Application) Nuclear counting statistics, shielding, and decay rate measurements.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.