Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Advanced Biology with Vernier
SDPSCI_ED

Catalog No. s16096nd Shop All Vernier Software Products
Click to view available options
:
17 ready-to-use lab experiments

Contains experiments appropriate for AP Biology and college biology courses.

  • Contains 17 ready-to-use lab experiments
  • Includes 12 recommended AP Biology labs with complete instructions for data collection
  • Can be used in college biology courses with several new experiments utilizing spectrometers

Specifications

For Use With (Application) Experiments appropriate for AP Biology and college biology courses
Product Type Lab Book
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.