Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Biology with Vernier
SDPSCI_ED

Catalog No. s16045nd Shop All Vernier Software Products
Click to view available options
:
Contains 31 experiments on important biology concepts

Foundational biology book for high school and college courses.

  • Contains 31 experiments in cell respiration, photosynthesis, membrane diffusion, osmosis, human physiology, transpiration, fermentation, and other important biology concepts
  • Experiments included for 12 Vernier biology sensors
  • Supports seven AP Biology lab activities
  • Includes materials list, step-by-step instructions, data tables, and questions
  • Contains Teacher Information section with directions for setting up experiments, helpful hints, and sample graphs and data
  • Word-processing files of student sections on CD

Specifications

For Use With (Application) Foundational biology book for high school and college courses
Product Type Lab Book
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.