Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Investigating Chemistry through Inquiry
SDPSCI_ED

Catalog No. s15961nd Shop All Vernier Software Products
Click to view available options
:
Contains 25 chemistry investigation activities

Contains chemistry investigations using inquiry method.

  • Includes 25 inquiry-based chemistry investigations
  • Topics comprise thermochemistry, acids and bases, stoichiometry, chemical kinetics, and properties of solutions
  • Authored by two chemistry teachers, Donald L. Volz and Ray Smola
  • Experiments include preliminary activity, teacher information, sample researchable questions, and sample data
  • Contains preliminary activities with step-by-step instructions, data tables, and questions
  • Teacher Information section with directions for setting up experiments, helpful hints, and sample graphs and data
  • Word-processing files of student sections on CD

Specifications

For Use With (Application) Learning chemistry investigations through inquiry
Product Type Lab Book
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.