Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Organic Chemistry with Vernier
SDPSCI_ED

Catalog No. s16120nd Shop All Vernier Software Products
Click to view available options
:
Contains experiments for use w/Vernier data-collection technology

Catalog No. S16120ND

Supplier: Vernier CHEMO

Only null left
Add to Cart
Add to Cart

Contains experiments for use with Vernier data-collection technology in college organic chemistry courses.

  • Experiments cover topics and techniques including compound identification, synthesis, chromatography, optical rotation, and spectroscopy
  • Helps integrate Vernier data-collection technology into organic chemistry lab curriculum
  • Experiments involve characterization of reactions and compounds using Vernier Melt Station, Mini Gas Chromatograph, SpectroVis Plus, and Polarimeter
  • Includes materials list, step-by-step instructions, data tables, questions
  • Teacher Information section with directions for setting up experiments, helpful hints, and sample graphs and data
  • Word-processing files of student sections on CD

Specifications

For Use With (Application) For use with Vernier data-collection technology
Product Type Lab Book
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.