Learn More
Vernier Standard Low Cl ISE Solution
SDPSCI_ED
Catalog No. s16124nd Shop All Vernier Software Products
Click to view available options
For Use With (Equipment):
Low Chloride ISE
Used to calibrate Ion-Selective Electrodes
  • 500mL solution has concentration of 10mg/L
Specifications
For Use With (Equipment) Low Chloride ISE
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.