Learn More
Vernier Standard Low Nitrate ISE Solution
SDPSCI_ED
Catalog No. s16219nd
Click to view available options
For Use With (Equipment):
Low nitrate ISE
Used to calibrate Ion-Selective Electrodes
  • 500mL solution has concentration of 1mg/L
Specifications
For Use With (Equipment) Low nitrate ISE
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.