Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VERO cells Rabbit anti-Monkey, Polyclonal, Abnova™

Catalog No. 89033669 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-669 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-669 Supplier Abnova Corporation Supplier No. PAB29824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against VERO cells.

  • High specificity
  • High reactivity

Specifications

Antigen VERO cells
Applications Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate FITC
Description Rabbit polyclonal antibody raised against VERO cells.
Dilution Flow Cytometry Immunofluorescence Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Formulation In Phosphate buffered saline, pH 7.4
Host Species Rabbit
Immunogen Vero-76 cells (ATCC CRL-1587).
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species African Green Monkey
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.