Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Versa-Clean™ Pharmaceutical Production and Multi-Purpose Cleaner
Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
1 L
4 L
Description
- Clean-out-of-place (COP): Manual labware washing, pharmaceutical washers, immersion soak tanks, ultrasonic cleaners. Clean-in-place (CIP): Mixing vessels, process equipment, piping, packaging equipment
- Use where glassware and sensitive lab equipment must be totally free of residues that could interfere with analysis, tissue culture, manufacturing and other procedures
- Third party lot specific tested for inhibitory residues to meet NELAC and State water lab audit criteria (Standard Methods for the Examination of Water and Wastewater 21st Ed., 9020B-4A-2)
- Does not contain: alcohol, ammonia, APE’s, caustic , chlorine, fragrance, glycol ether solvents, NPE’s, NTA, phosphates, silicates, solvents, sulfates, volatile organic compounds or ozone depleting substances
- Readily biodegradable in 7 days (OECD 301D) and phosphate free
- Mild pH of 9.75 it can be disposed of straight to the drain
- Complete solubility, works equally well in hot or cold water
- Formulated for controlled foaming
- Fisherbrand Versa-Clean is manufactured using sustainable, low waste and low energy processes in an audited, modern plant, using state of the art, precision production equipment
Order Info
Third-party lot specific tested for inhibitory residues to meet NELAC and State water lab audit criteria.
Certifications
Passes inhibitory residue test.
Specifications
Specifications
| Form | Liquid |
| For Use With (Application) | For use in ultrasonic cleaners, high-pressure sprayers, scrubbers/soak tanks |
| Volume (Metric) | 4 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction