Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Versa-Clean™ Pharmaceutical Production and Multi-Purpose Cleaner
SCI_ED

Catalog No. p-6707022 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
1 L
4 L

Fisherbrand Versa-Clean™ From delicate labware to full scale production equipment Versa-Clean keeps sensitive equipment free of inhibitory residues that could interfere with analysis, tissue culture, and other critical procedures. Ideal for pharmaceutical production, research labs, environmental labs, water or wastewater labs.

  • Clean-out-of-place (COP): Manual labware washing, pharmaceutical washers, immersion soak tanks, ultrasonic cleaners. Clean-in-place (CIP): Mixing vessels, process equipment, piping, packaging equipment
  • Use where glassware and sensitive lab equipment must be totally free of residues that could interfere with analysis, tissue culture, manufacturing and other procedures
  • Third party lot specific tested for inhibitory residues to meet NELAC and State water lab audit criteria (Standard Methods for the Examination of Water and Wastewater 21st Ed., 9020B-4A-2)
  • Does not contain: alcohol, ammonia, APE’s, caustic , chlorine, fragrance, glycol ether solvents, NPE’s, NTA, phosphates, silicates, solvents, sulfates, volatile organic compounds or ozone depleting substances
  • Readily biodegradable in 7 days (OECD 301D) and phosphate free
  • Mild pH of 9.75 it can be disposed of straight to the drain
  • Complete solubility, works equally well in hot or cold water
  • Formulated for controlled foaming
  • Fisherbrand Versa-Clean is manufactured using sustainable, low waste and low energy processes in an audited, modern plant, using state of the art, precision production equipment

Certifications

Passes inhibitory residue test.

Specifications

Form Liquid
pH 9.75 to 10
For Use With (Application) For use in ultrasonic cleaners, high-pressure sprayers, scrubbers/soak tanks
Quantity 12 Pk.
Volume (Metric) 1 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.